Learn More
Brady™ Polyester Label Cartridges for BMP™21-PLUS and BMP™21-LAB Printers
Polyester Label Cartridges for use with BMP™21-PLUS and BMP™21-LAB printers.
248.00 DKK - 454.00 DKK
Specifications
Product Type | Label Cartridge |
---|---|
Print Technology | Thermal Transfer |
Length (English) | 21 ft. |
Length (Metric) | 6.4 m |
Product Code | Brand | Color | For Use With (Equipment) | Material | Width (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | For Use With (Equipment) | Material | Width (Metric) | Price | Quantity & Availability | |||
11844345
|
Brady™
110896 |
White | Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer, BMP21-LAB Label Printer, BMP21-PLUS Label Printer | Polyester | 9.53 mm |
454.00 DKK
Each |
In Stock
Log in to see stock. |
|||
11830835
|
Brady™
110899 |
Clear | Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer, BMP21-LAB Label Printer, BMP21-PLUS Label Printer | Polyester | 9.53 mm |
268.00 DKK
Each |
In Stock
Log in to see stock. |
|||
11593864
|
Brady™
110900 |
Clear | Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer, BMP21-LAB Label Printer, BMP21-PLUS Label Printer | Polyester | 12.70 mm |
368.00 DKK
Each |
Estimated Shipment: 08-08-2025
Log in to see stock. |
|||
11890875
|
Brady™
110901 |
Clear | Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer, BMP21-LAB Label Printer, BMP21-PLUS Label Printer | Polyester | 19.05 mm |
350.00 DKK
Each |
In Stock
Log in to see stock. |
|||
11820835
|
Brady™
110898 |
White | Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer, BMP21-LAB Label Printer, BMP21-PLUS Label Printer | Polyester | 19.05 mm |
429.00 DKK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||
15608454
|
Brady™
139755 |
Clear | Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer, BMP21-LAB Label Printer, BMP21-PLUS Label Printer | Polyester (B-430) | 6.35 mm |
304.00 DKK
Each |
In Stock
Log in to see stock. |
|||
17851485
|
Brady™
139746 |
Clear | Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer, BMP21-LAB Label Printer, BMP21-PLUS Label Printer | Polyester (B-430) | 6.35 mm |
248.00 DKK
Each |
In Stock
Log in to see stock. |
|||
17881485
|
Brady™
139747 |
White | Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer, BMP21-LAB Label Printer, BMP21 Label Printer, BMP21-PLUS Label Printer, IDPAL Handheld Label Maker, LABPAL Handheld Label Maker | Polyester (B-430) | 12.70 mm |
300.00 DKK
Each |
In Stock
Log in to see stock. |
|||
17821505
|
Brady™
139748 |
Clear | Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer, BMP21-LAB Label Printer, BMP21 Label Printer, BMP21-PLUS Label Printer, IDPAL Handheld Label Maker, LABPAL Handheld Label Maker | Polyester (B-430) | 9.53 mm |
268.00 DKK
Each |
In Stock
Log in to see stock. |
|||
15509405
|
Brady™
139749 |
Clear | Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer, BMP21-LAB Label Printer, BMP21 Label Printer, BMP21-PLUS Label Printer, IDPAL Handheld Label Maker, LABPAL Handheld Label Maker | Polyester (B-430) | 19.05 mm |
429.00 DKK
Each |
In Stock
Log in to see stock. |
|||
Description
- For smooth surface indoor labeling
- Made of white or clear self laminating polyester material
- Glossy finish and permanent acrylic adhesive
- Compatible with Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer, BMP21 LAB Label Printer, BMP21 Label Printer, BMP21-PLUS Label Printer, IDPAL Handheld Label Maker, LABPAL Handheld Label Maker
- M21-250-423, M21-250-430 and M21-250-430-WT-CL are compatible only with the BMP™21-PLUS printer
Applications:
Asset Tracking, Barcoding, Component ID, Equipment ID, General ID, General Identification, General Purpose ID, Location ID, Outlet/Jack ID, Patch Panel, Patch Panel - ID Port, Plate, Rating Plates, DataComm Equipment
Compliance
RoHS Compliant, UL Recognized, CSA Approved, AGA Approved, Halogen-Free (DIN VDE 0472/parts 815)
Specifications
Label Cartridge | |
Thermal Transfer | |
21 ft. | |
6.4 m |