Learn More
BRAND™ Titrette™ Digital Bottle-Top Burettes
Provides accuracy and precision within the tolerances of Class A glass burettes. Titrette™ Digital Bottle-top Burets are suitable for most titrants in concentrations up to 1 mol/L. Ideal for general chemistry, wastewater treatment applications, food/beverage analysis, industrial titrations, environmental fieldwork, and more.
8725.00 DKK - 11455.00 DKK
Specifications
Product Type | Digital Bottletop Dispenser |
---|---|
Angle | 90° |
Autoclavable | Not Autoclavable |
Certifications/Compliance | DIN EN ISO 385, ASTM E287 |
Color | Gray |
Product Code | Brand | Volume (Metric) | Interface | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Interface | Price | Quantity & Availability | |||||
10703164
![]() |
BRAND™
4760151 |
25 mL | RS232 Interface |
8920.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
10703354
![]() |
BRAND™
4760161 |
50 mL | RS232 Interface |
8725.00 DKK
Each |
Estimated Shipment: 08-08-2025
Log in to see stock. |
|||||
10687425
![]() |
BRAND™
4760251 |
25 mL | Without Interface |
11455.00 DKK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
10769165
![]() |
BRAND™
4760261 |
50 mL | Without Interface |
11455.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
15394127
![]() |
BRAND™
4760141 |
10 mL | RS232 Interface |
8920.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
15604077
![]() |
BRAND™
4760241 |
10 mL | Without Interface |
11455.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
Description
Provides accuracy and precision within the tolerances of Class A glass burettes. BrandTech™ BRAND™ Titrette™ Digital Bottle-top Burets are suitable for most titrants in concentrations up to 1 mol/L. Ideal for general chemistry, wastewater treatment applications, food/beverage analysis, industrial titrations, environmental fieldwork, and more.
The Titrette™ bottle-top burette provides accuracy and precision within the tolerances of Class A glass burettes. Suitable for most titrants in concentrations up to 1 mol/L, it is ideal for general chemistry, wastewater treatment applications, food/beverage analysis, industrial titrations, environmental fieldwork, and more. The Titrette will enable you to perform routine titrations quickly, accurately, and with ease.
- Smooth operation for exceptionally sensitive drop-wise titrations
- Avoids the risk of spills from poured transfers
- Eliminates meniscus reading errors
- Switch automatically between filling and titrating
- Meets tolerances for Class A glass burettes per ISO and ASTM standards
- Optional RS-232 interface available for direct data transmission to a PC
- Use for a variety of applications including general chemistry, water treatment, food/beverage analysis, industrial titrations, and environmental work in the lab or field
Order Info
For burettes with Calibration Certificate, contact Customer Service. Add DKD at the end of the BRANDTECH part number. No returns accepted.
Specifications
Digital Bottletop Dispenser | |
Not Autoclavable | |
Gray | |
Digital | |
15°C to 40°C | |
1.5 V |
90° | |
DIN EN ISO 385, ASTM E287 | |
Glass | |
59°F to 104°F | |
Up to 500 mm 2/s |