Learn More
BrandTech™ PFA Sample Containers
VITLAB PFA sample containers supplied with screw cap and are suitable for trace analysis work.
442.00 DKK - 487.00 DKK
Specifications
Product Type | Sample Container |
---|---|
Material | PFA |
For Use With (Application) | Sample Collection, Transport and Storage |
Product Code | Brand | Capacity | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Capacity (Metric) | Price | Quantity & Availability | |||||
10060641
|
BrandTech™
130397 |
60 mL | 60 mL |
442.00 DKK
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
|||||
10070641
|
BrandTech™
130497 |
90 mL | 90 mL |
487.00 DKK
Each |
Estimated Shipment: 18-06-2025
Log in to see stock. |
|||||
Description
- Suitable for sample collection, transport and storage.
- Autoclavable at 121 C
Specifications
Sample Container | |
PFA | |
Sample Collection, Transport and Storage |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BrandTech™ PFA Sample Containers