Learn More
BRAND™ Polypropylene Thread Adapters for Bottle-Top Dispensers
Adapt current model Dispensette™ seripettor™, Titrette™ and QuikSip™ to common bottle sizes.
Brand: BRAND™ 704340
73.86 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Polypropylene Construction
- For use with Dispensette™ seripettor™ Titrette™ and QuikSip™ Bottle-top dispensers
Specifications
Bottle Thread Adapter | |
Dispensette™, seripettor™, Titrette™ and QuikSip™ | |
GL 32/S 40 |
Polypropylene | |
1 Ea. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
BRAND™ Polypropylene Thread Adapters for Bottle-Top Dispensers > For S 40 bottle thread
Spot an opportunity for improvement?Share a Content Correction