Learn More
BrandTech™ PFA Sample Tubes
For sample preparation, centrifugation and autosampling
Brand:  BrandTech™ 103897
Product Code. 10320361
  • 554.00 DKK / Each
Only null left
Estimated Shipment: 01-09-2025
Specifications
Autoclavable
PFA
Sample preparation, centrifugation, and autosampler racks for trace analysis
PFA
110 mm
15 mL
GL25
Yes
0.5 oz.
Screw Cap
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BrandTech™ PFA Sample Tubes > 15mL