missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Single-Channel Micropipette Transferpette™ Electronic, DE-M
Ergonomic, handy design. Clear and intuitive operation. Up to 4,000 pipetting cycles on a single charge.
Brand: BRAND™ 705323
This item is not returnable.
View return policy
Description
- Transferpette electronic pipettes combine the globally proven properties of mechanical piston operated air displacement pipettes by BRAND with the advantages of an electronic device
- The pipette shaft can be unscrewed and is entirely autoclavable (121 °C).
- Five convenient programs: pipetting, reverse pipetting, mixing, GEL-Electrophoresis and dispensing.
- Easy Calibration-Technique for making quick adjustments of the Transferpette™ electronic, without tools.
- Limitations of use:<br />Operating temperature of instrument and reagent should be between +15 °C and +40 °C (59 °F to 104 °F) (other temperatures upon request)
- Vapor pressure up to 500 mbar
- Viscosity: 260 mPa s
- For viscous media, the speed must be adjusted if necessary.

Specifications
≤ ± 0.8%, 1.6μL | |
DIN EN ISO 8655-2 | |
Yellow | |
Transferpette | |
10 to 200 μL | |
Ergonomic | |
0.2μL | |
59°F to 104°F | |
1 Year |
Rechargeable NiMH | |
Autoclavable | |
1 | |
Singel Channel Microliter Pipette | |
LCD | |
USA/Japan AC Adapter | |
1 | |
15°C to 40°C | |
110 V/50-60 Hz |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ Single-Channel Micropipette Transferpette™ Electronic, DE-M > 10 to 200 uL; With AC charger for USA/Japan
Spot an opportunity for improvement?Share a Content Correction