Learn More
VACUUBRAND™ Service Kits for NT Series Chemistry Pumps and Systems
Service kits for VACUUBRAND NT series Chemistry design pumps.
Brand: VACUUBRAND™
1401.25 DKK valid until 2025-08-24
Use promo code "24140" to get your promotional price.
This item is not returnable.
View return policy
Description
- Sets include appropriate diaphragms, valves and sealing rings (if applicable)
- Wrench not included.
Specifications
MZ 2C NT, MZ2C NT VARIO, PC201 NT, PC3002 VARIO | |
Service Kit |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59%
Product Content Correction
Product Title
VACUUBRAND™ Service Kits for NT Series Chemistry Pumps and Systems > For MZ2C NT, MZ2C NT VARIO, PC201 NT, PC3002 VARIO
Spot an opportunity for improvement?Share a Content Correction