Learn More
R&L Enterprises™ Cork Borer
Efficiently and cleanly bores holes in cork. R&L Enterprises™ Cork Borers have a cutting tube mounted with safety under-flange in shaped handle.
Brand:  R&L Enterprises™ 3402
 View more versions of this product
Product Code. 12873952
  • 2005.00 DKK / Each
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Efficiently and cleanly bores holes in cork. R&L Enterprises™ Cork Borers have a cutting tube mounted with safety under-flange in shaped handle.
  • With rod for clearning bores
  • Manufactured from brass
  • Sizes numbered consecutively from 1 upwards in each set
Specifications
Specifications
Range: 4 to 25mm
Cork Borer
Brass
Product Title
R&L Enterprises™ Cork Borer > Range: 4 to 25mm