missing translation for 'onlineSavingsMsg'
Learn More

National Lab Bravo Grid Divider, Cardboard

For organizing, storing, and archiving samples in cryoboxes.

Brand:  National Lab RB10650505N

 View more versions of this product

Product Code. 12400671

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Cardboard construction
  • Suitable for dry and refrigerated storage
  • Suitable for sample archiving
  • Suitable for cryoboxes
Specifications

Specifications

5x5
Grid Divider
Cryoboxes 133 x 133 mm
65 mm
Fiberboard
Dry and Refrigerated Storage/Archiving
25 (24.5 mm Diameter Tubes)
24.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.