missing translation for 'onlineSavingsMsg'
Learn More
Learn More
National Lab Bravo Grid Divider, Cardboard
For organizing, storing, and archiving samples in cryoboxes.
Brand: National Lab RB10300808N
This item is not returnable.
View return policy
Description
- Cardboard construction
- Suitable for dry and refrigerated storage
- Suitable for sample archiving
- Suitable for cryoboxes
Specifications
8x8 | |
Grid Divider | |
Cryoboxes 133 x 133 mm | |
30 mm | |
15 mm |
Fiberboard | |
Dry and Refrigerated Storage/Archiving | |
64 (15 mm Diameter Tubes) | |
15 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction