Learn More
Invitrogen™ BrightComp eBeads™ Compensation Bead Kits
BrightComp eBeads compensation beads provide a consistent, accurate, and simple-to-use technique for setting flow cytometry compensation when using fluorescent protein-expressing samples.
Brand:  Invitrogen™ A54741
1360.10 DKK valid until 2025-06-30
Use promo code "24074" to get your promotional price.
 View more versions of this product
Product Code. 17807071
  • 2030.00 DKK / Pack of 25
Explore more special offers
Description
Description
BrightComp eBeads compensation beads provide a consistent, accurate, and simple-to-use technique for setting flow cytometry compensation when using fluorescent protein-expressing samples.
• Three levels of intensity, perfect for any samples no matter the level of expression
• Near-identical, spectrally-matched dye to fluorescent protein
• Consistent, reliable performance
Accurate compensation is crucial for effective multicolor analysis in flow cytometry. Each kit provides polystyrene microspheres with near-identical, spectrally-matched fluorescent protein dye as well as negative control beads in a single dropper bottle.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
YFP BrightComp eBeads Compensation Bead Kit
25 tests
Compensation Beads
488 nm
Flow Cytometry
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3091990 Certificate TypeCertificate of Analysis Date18/11/2024 Catalog NumberA54741,A54741_CLEAN
Lot Number2876750 Certificate TypeCertificate of Analysis Date26/03/2024 Catalog NumberA54741,A54741_CLEAN
Lot Number2738314 Certificate TypeCertificate of Analysis Date29/02/2024 Catalog NumberA54741,A54741_CLEAN
Lot Number2729785 Certificate TypeCertificate of Analysis Date14/07/2023 Catalog NumberA54741,A54741_CLEAN
Lot Number2706266 Certificate TypeCertificate of Analysis Date11/05/2023 Catalog NumberA54741,A54741_CLEAN
Product Title
Invitrogen™ BrightComp eBeads™ Compensation Bead Kits > YFP BrightComp eBeads™ Compensation Bead Kit