missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromide Standard for Ion Chromatography, 0.1M NaBr, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
460.00 DKK - 890.00 DKK
Chemical Identifiers
CAS | 7647-15-6, 7732-18-5 |
---|---|
Molecular Formula | BrNa |
Molecular Weight (g/mol) | 102.89 |
MDL Number | MFCD00003475 |
InChI Key | JHJLBTNAGRQEKS-UHFFFAOYSA-M |
Synonym | sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a |
PubChem CID | 253881 |
ChEBI | CHEBI:63004 |
IUPAC Name | sodium bromide |
SMILES | [Na+].[Br-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
16361136
|
SPEX Certiprep
AS-BR9-5Y |
125 mL | Triple-leached LDPE bottle |
460.00 DKK
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16351136
|
SPEX Certiprep
AS-BR9-5X |
500 mL | Triple-leached LDPE bottle |
890.00 DKK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Br- | 0.1M |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7647-15-6, 7732-18-5 | |
102.89 | |
JHJLBTNAGRQEKS-UHFFFAOYSA-M | |
253881 | |
sodium bromide |
BrNa | |
MFCD00003475 | |
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a | |
CHEBI:63004 | |
[Na+].[Br-] |
Specifications
7647-15-6, 7732-18-5 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00003475 | |
JHJLBTNAGRQEKS-UHFFFAOYSA-M | |
sodium bromide | |
253881 | |
0.1M NaBr | |
Bromide Standard |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
BrNa | |
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a | |
[Na+].[Br-] | |
102.89 | |
CHEBI:63004 | |
H2O |
Safety and Handling
missing translation for 'dotInformation' : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only