Learn More
Bromoethane, 98%, Thermo Scientific Chemicals
Description
Bromoethane is used in organic synthesis for the introduction of ethyl group. It is widely used as solvent, as an anesthetic in medicine and as a refrigerant. It is an important component of antiknock fluid in gasoline. It is also also used in the manufacture of pharmaceuticals like vitamin A as well as in the manufacture of flame retardants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Bromoethane is used in organic synthesis for the introduction of ethyl group. It is widely used as solvent, as an anesthetic in medicine and as a refrigerant. It is an important component of antiknock fluid in gasoline. It is also also used in the manufacture of pharmaceuticals like vitamin A as well as in the manufacture of flame retardants.
Solubility
Miscible with ethanol, ether, chloroform and organic solvents.
Notes
Light sensitive. Store in cool place. Incompatible with strong bases, strong oxidizing agents, magnesium, potassium, calcium, sodium oxides, aluminum, zinc and alkali metals.

Specifications
Specifications
Melting Point | -119°C |
Density | 1.461 |
Boiling Point | 37°C to 39°C |
Flash Point | −23°C (−9°F) |
Assay Percent Range | ≥98% |
Odor | Ether-like |
Linear Formula | BrCH2CH3 |
Refractive Index | 1.4235 |
Quantity | 100 g |
UN Number | UN1891 |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.