Learn More
Thermo Scientific Chemicals Bromothymol Blue Product Code.: 11423348
Thermo Scientific Chemicals Bromothymol Blue
Product Code. 11423348
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
Product Code. 11423348
Brand: Thermo Scientific Chemicals A17746.18
Request Bulk or Custom Format
Bromothymol Blue is a pH indicator used for measuring pH around 7 in pools and fish tanks. It operates in the pH range 6.0 to 7.6 due to the presence of an electron withdrawing and donating groups like bromine atoms and alkyl substitutents respectively. In the laboratory, it is used as a biological slide stain as well as used in obstetrics for detecting premature rupture of membranes. Further, it is used for the observation of photosynthetic activities and a respiratory indicator. In addition to this, it is used to define cell walls or nuclei under the microscope.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bromothymol Blue is a pH indicator used for measuring pH around 7 in pools and fish tanks. It operates in the pH range 6.0 to 7.6 due to the presence of an electron withdrawing and donating groups like bromine atoms and alkyl substitutents respectively. In the laboratory, it is used as a biological slide stain as well as used in obstetrics for detecting premature rupture of membranes. Further, it is used for the observation of photosynthetic activities and a respiratory indicator. In addition to this, it is used to define cell walls or nuclei under the microscope.
Solubility
Soluble in ethanol, ether and alkalis. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 76-59-5
Molecular Formula C27H28Br2O5S
Molecular Weight (g/mol) 624.384
MDL Number MFCD00005872
InChI Key NUHCTOLBWMJMLX-UHFFFAOYSA-N
Synonym Bromthymol Blue
PubChem CID 6450
ChEBI CHEBI:86155
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenolShow MoreShow Less
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br
Specifications
Melting Point 200°C to 202°C
Beilstein 373934
Merck Index 14,1445
Solubility Information Soluble in ethanol,ether and alkalis. Slightly soluble in water.
Formula Weight 624.39
Quantity 50 g
Chemical Name or Material Bromothymol Blue
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249282 Certificate TypeCertificate of Analysis Date24/01/2024 Catalog NumberA17746.18,A17746.09,A17746.30
Lot Number10242336 Certificate TypeCertificate of Analysis Date28/07/2022 Catalog NumberA17746.18,A17746.09,A17746.30
Lot Number10237337 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA17746.18,A17746.09,A17746.30
Lot Number10122869 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17746.18,A17746.09,A17746.30
Lot Number10172474 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17746.18,A17746.09,A17746.30
RUO – Research Use Only
Product Content Correction
Product Title
Thermo Scientific Chemicals Bromothymol Blue >