Learn More
Bronze powder, -100 mesh, 99.0% (metals basis)
Description
Bronze powder suitable for a range of applications including resin-casting, decorative coatings and powder metallurgy. Also used for hydraulic seals, surface engineering, decorative, welding and brazing, diamond tooling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Chemical Name or Material | Bronze |
Appearance | Bronze colored powder |
Assay Percent Range | 99% (Metals basis) |
MDL Number | MFCD00198187 |
Quantity | 250 g |
Solubility Information | Insoluble in water. |
Mesh Size | -100 Mesh |
Odor | Odorless |
Form | Powder |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
W24A039 | Certificate of Analysis | 15/01/2025 | 088359.30,088359.A1 |
Y09I035 | Certificate of Analysis | 18/11/2022 | 088359.30,088359.A1 |
W06C010 | Certificate of Analysis | 08/12/2021 | 088359.30,088359.A1 |
K05P10 | Certificate of Analysis | 08/12/2021 | 088359.30,088359.A1 |
F26M37 | Certificate of Analysis | 08/12/2021 | 088359.30,088359.A1 |