Learn More
Thermo Scientific™ Buckets and Lids for Thermo Scientific™ TX-150 Swinging Bucket Rotor
Round and conical buckets offered in sets of four
Brand: Thermo Scientific™ 75005702
5563.30 DKK valid until 2025-07-04
Use promo code "24147" to get your promotional price.
Description
Round and conical buckets offered in sets of four
Specifications
4 x 145 mL | |
Bucket | |
Set of 4 | |
Round Buckets |
Promotions
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30
-
Multiple Manufacturers
Cancer Research Campaign
Save big on essential products for cancer researchPromo Code: 24147Expires: 2025-07-31
New Offer Save Now -
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59% -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13