Learn More
Buffer Concentrated Solution pH 4 (Phthalate)
Shop All Fisher Chemical Products
Click to view available options
Quantity:
6 Ampules
Unit Size:
6 ampoules
Specifications
Specifications
Color | Colorless |
pH | 4 |
Physical Form | Liquid |
Quantity | 6 Ampules |
MDL Number | 217672 |
Packaging | Ampoule |
Documents
Scientific Resources
Brochures
Fisherbrand Focus on pH and electrochemistry
File size: 9.1 MB
Fisherbrand Focus on pH and electrochemistry
File size: 2.5 MB
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
2539423 | Certificate of Analysis | 09/06/2025 | J/2820/17,J/2820/08,J/2820C/90,J/2820/24,J/2820/15,J/2820/21 |
2537787 | Certificate of Analysis | 24/05/2025 | J/2820/17,J/2820/08,J/2820C/90,J/2820/24,J/2820/15,J/2820/21 |
2531600 | Certificate of Analysis | 24/05/2025 | J/2820/17,J/2820/08,J/2820C/90,J/2820/24,J/2820/15,J/2820/21 |
2529546 | Certificate of Analysis | 24/05/2025 | J/2820/17,J/2820/08,J/2820C/90,J/2820/24,J/2820/15,J/2820/21 |
Promotions
-
Multiple Manufacturers
Science Is Hard Enough Your Bioreagents Shouldn’t Be
Reliable performance. Special prices.Happier researchers.Promo Code: 24128Expires: 2025-08-31
New Offer Save Now
Product Content Correction
Product Title
Buffer Concentrated Solution pH 4 (Phthalate) >
Spot an opportunity for improvement?Share a Content Correction