missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 7, Scharlab
Click to view available options
Quantity:
250 mL
1 L
Description
Standard buffer solutions are prepared using gravimetric and volumetric procedures. The batch value is determined by measurement with a combination glass electrode against five-point calibration according to DIN 19268. This pH buffer solution is traceable to Standard Reference Material from NIST.
Specifications
Specifications
Melting Point | -5°C |
Boiling Point | 109°C |
Density | 1.01g/cm3 |
pH | 7.00 |
Physical Form | Solution |
Quantity | 250 mL |
Formulation | Per liter: 3.54g Potassium dihydrogen phosphate, 14.7g Disodium hydrogen phosphate. Contains preservative. |
Traceability to NIST | Traceable to NIST |
Solubility Information | Miscible in water (20°C) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'