missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 9.00 (20°C), Fluka™

Certified, colored yellow

Brand:  Honeywell-Fluka 35082-500ML

Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 16222892

  • 316.27 DKK / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Buffer Solution pH 9.00 (20°C)
<0°C
Yellow
500 mL
12179-04-3, 10043-35-3
100°C (1.013 hPa)
9
Plastic Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buffer Solution pH 9.00 (20°C), Fluka™ > Quantity: 500 mL; Packaging: Plastic Bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.