Learn More
Wld Tec™ Flame 100 Safety Bunsen Burner
Safe alternative for traditional Bunsen or alcohol burners. Wld Tec™ Flame 100 Safety Bunsen Burner is ideally suited for all flame-related laboratory applications.
Brand:  Wld Tec™ 2.100.000
Product Code. 11774169
  • 2925.00 DKK / Each
Only null left
Estimated Shipment: 22-08-2025
Description
  • Multi-use
    • Sutiable for all flame-related laboratory applications
    • Precison flame allows graduated heating as well as safe sterilization
    • Flame size and intensity can be infinitely adjusted
  • Easy operation
    • Activated with the push of a button
    • Optional foot pedal control
    • Optional exernal IR-Sensor control
  • Reliable safety
    • Safety control system (SCS)
    • Gas shut-off activation
    • Overheating protection
    • Burner head clogging detection
  • Efficient
    • Economical to operate
    • Minimized gas consumption
  • Compact and sturdy
    • Stainless steel housing and burner
    • Removable burner tube for easy claining
    • Small footprint
Specifications
Specifications
Laboratory Gas Burner
Natural Gas, Butane, Propane
34 mm
1 Ea.
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Wld Tec™ Flame 100 Safety Bunsen Burner > Height: 34mm