Learn More
Usbeck Laborgeraete™ Bunsen Burner
234.00 DKK - 402.00 DKK
Specifications
Product Type Bunsen Burner
No. per Pack 1 Ea.
View More Specs
Products 6
Product Code Brand Height (Metric) Max. Temperature (Metric) Gas Type Price Quantity & Availability  
Product Code Brand Height (Metric) Max. Temperature (Metric) Gas Type Price Quantity & Availability  
10692351
View Documents
Usbeck Laborgeraete™
1020
145 mm 1500°C Natural Gas
309.52 DKK
Each
Only null left
Estimated Shipment: 05-08-2025
Add to basket
10447231
View Documents
Usbeck Laborgeraete™
1040/1
150 mm 1580°C Natural Gas
402.00 DKK
Each
Only null left
Estimated Shipment: 29-07-2025
Add to basket
10653511
View Documents
Usbeck Laborgeraete™
1010
145 mm 1540°C Natural Gas
254.33 DKK
Each
Only null left
Estimated Shipment: 29-07-2025
Add to basket
10413691
View Documents
Usbeck Laborgeraete™
1021
145 mm 1550°C Propane
352.00 DKK
Each
Only null left
Estimated Shipment: 05-08-2025
Add to basket
10618562
View Documents
Usbeck Laborgeraete™
1000
145 mm 1530°C Natural Gas
234.00 DKK
Each
Only null left
Estimated Shipment: 05-08-2025
Add to basket
10330372
View Documents
Usbeck Laborgeraete™
1001
145 mm 1580°C Propane
234.00 DKK
Each
Only null left
Estimated Shipment: 05-08-2025
Add to basket
Description
Description
Specifications
Specifications
Bunsen Burner
1 Ea.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Usbeck Laborgeraete™ Bunsen Burner