Learn More
Karl Hecht™ Assistent™ Burette Tips Product Code.: 10217521
Only undefined left
Add to basket
Karl Hecht™ Assistent™ Burette Tips
Product Code. 10217521
Burette tips with rubber tubing for burettes with tubing connection
Click to view available options
:
Burette tips with rubber tubing for burettes with tubing connection
This item is not returnable. View return policy
Product Code. 10217521
Brand: Karl Hecht™ 40780009
Only undefined left
Add to basket
Only undefined left
Add to basket
This item is not returnable. View return policy
Excellent for dispensing measured amounts of chemical solutions in the laboratory. Karl Hecht™ Assistent™ Burette Tips are manufactured from AR-Glas™ soda glass with rubber tubing.
  • Length approx. 60mm
  • For burettes with tubing connection
Specifications
For Use With (Equipment) Burets with tubing connection
Includes Rubber Tubing
Type Buret Tip
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Karl Hecht™ Assistent™ Burette Tips >