missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C4BPA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11819-1-AP-20UL
This item is not returnable.
View return policy
Description
C4BP is composed of two polypeptides (alpha and beta chains), which have molecular weights of 70 and 45 kDa, respectively. This polyclonal antibody is raised against the C4BP alpha chain (C4BPA).
This gene encodes a member of a superfamily of proteins composed predomitly of tandemly arrayed short consensus repeats of approximately 60 amino acids. Along with a single, unique beta-chain, seven identical alpha-chains encoded by this gene assemble into the predomit isoform of C4b-binding protein, a multimeric protein that controls activation of the complement cascade through the classical pathway. The genes encoding both alpha and beta chains are located adjacent to each other on human chromosome 1 in the regulator of complement activation gene cluster. Two pseudogenes of this gene are also found in the cluster.Specifications
C4BPA | |
Polyclonal | |
Unconjugated | |
C4BPA | |
C4BP, C4BPA, Proline rich protein, PRP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
722 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04003 | |
C4BPA | |
C4BPA Fusion Protein Ag2387 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C4BPA Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction