missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CABP5 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-06122AF405
This item is not returnable.
View return policy
Description
CABP5 Polyclonal specifically detects CABP5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CABP5 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CABP3, calcium binding protein 5, calcium-binding protein 3, calcium-binding protein 5 | |
Produced in rabbits immunized with E. coli-derived Human CABP5 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
56344 | |
Store at 4°C in the dark. | |
IgG |