Learn More
Cadmium shot, tear drop, -5+20 mesh, 99.95% (metals basis) Product Code.: 11371168
Cadmium shot, tear drop, -5+20 mesh, 99.95% (metals basis)
Product Code. 11371168
50 g, 50g
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 11371168
Brand: Thermo Scientific Alfa Aesar 010122.18
Request Bulk or Custom Format
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
CAS: 7440-43-9 | Cd | 112.41 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
PubChem CID 23973
ChEBI CHEBI:22977
IUPAC Name cadmium
SMILES [Cd]
Specifications
Quantity 50 g
Percent Purity 99.95%
Mesh Size −5+20 Mesh
Form Shot
Assay (metals basis)
Chemical Name or Material Cadmium shot
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberR23K029 Certificate TypeCertificate of Analysis Date11/06/2024 Product Code010122.A1,010122.30,010122.18
Lot NumberT27E035 Certificate TypeCertificate of Analysis Date23/04/2024 Product Code010122.A1,010122.30,010122.18
Lot NumberP21I048 Certificate TypeCertificate of Analysis Date12/04/2022 Product Code010122.A1,010127.95,010122.30,010122.18
Lot NumberL21Y048 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code010122.A1,010127.95,010122.30,010122.18
Lot NumberY27D006 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code010122.A1,010127.95,010122.30,010122.18
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cadmium shot
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P201-Obtain special instructions before use.
  • P320-Specific treatment is urgent (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
RUO – Research Use Only
Product Content Correction
Product Title
Cadmium shot, tear drop, -5+20 mesh, 99.95% (metals basis) >