missing translation for 'onlineSavingsMsg'
Learn More
Calcein, MP Biomedicals™ Product Code.: 11464622

Calcein, MP Biomedicals™

Product Code. 11464622 Shop All MP Biomedical Products
10 g
Click to view available options
Quantity:
10 g
25 g
This item is not returnable. View return policy

Product Code. 11464622

Brand: MP Biomedicals 0219016710

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1461-15-0
Molecular Formula C30H26N2O13
Molecular Weight (g/mol) 622.539
InChI Key DEGAKNSWVGKMLS-UHFFFAOYSA-N
Synonym Fluorexon, Calcein W
PubChem CID 65079
ChEBI CHEBI:51903
IUPAC Name 2-[[7'-[[bis(carboxymethyl)amino]methyl]-3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-2'-yl]methyl-(carboxymethyl)amino]acetic acid
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)CN(CC(=O)O)CC(=O)O)O)O)CN(CC(=O)O)CC(=O)O

Specifications

Formula Weight 622.5
Color Orange
Quantity 10 g
Physical Form Powder
Chemical Name or Material Calcein, Suitable for fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcein, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.