missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride Dihydrate, ACS Reagent Grade, Scharlab
Click to view available options
Quantity:
1 kg
Description
- Molecular Formula: CaCl2 • 2H2O
- Molar Mass: 147.02g/mol
- CAS: 10035-04-8
- EIN ECS No.: 233-140-8
- Oxidizing Substances (as NO3): 0.003% max
Specifications
Specifications
Melting Point | ∼176°C |
CAS | 10035-04-8 |
pH | 4.5–8.5 (5%, water) |
Physical Form | Powder |
Quantity | 1 kg |
Molecular Formula | CaCl2 • 2H2O |
Formula Weight | 147.02g/mol |
Percent Purity | 99 to 105% (complexometric) |
Grade | ACS Reagent |
Identification | Passes Test |
Safety and Handling

Product Identifier
- Calcium chloride dihydrate, powder, reagent grade, ACS
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Calcium Chloride Dihydrate, ACS Reagent Grade, Scharlab >
Spot an opportunity for improvement?Share a Content Correction