missing translation for 'onlineSavingsMsg'
Learn More
Calcium Hydroxide, 90%, Synthesis Grade, Scharlab Product Code.: 11354204

Calcium Hydroxide, 90%, Synthesis Grade, Scharlab

Product Code. 11354204
1 kg, 1kg
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
This item is not returnable. View return policy

Product Code. 11354204

Brand: Scharlab CA02151000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in building materials, in lubricant compositions, in painting and as a laboratory reagent.

  • Molecular Formula: Ca(OH)2
  • Molar Mass: 74.09g/mol
  • CAS: 1305-62-0
  • EIN ECS No.: 215-137-3

Specifications

CAS 1305-62-0
Quantity 1 kg
Molecular Formula CaH2O2
MDL Number MFCD00010901
Synonym calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
SMILES [OH-].[OH-].[Ca++]
IUPAC Name calcium dihydroxide
Molecular Weight (g/mol) 74.09
PubChem CID 6093208
ChEBI CHEBI:31341
Formula Weight 74.09g/mol
Percent Purity ∼90% (acidimetric)
Grade Synthesis
Show More Show Less
compliance-icons
Product Identifier
  • Calcium hydroxide, 90%, synthesis grade
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium Hydroxide, 90%, Synthesis Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.