Learn More
Calmodulin 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5121166
Description
Positive test controls include: NIH/3T3, MCF7, BT-474, NCI-H460, Mouse brain. The target is usually found in the following locations: Cytoplasm, cytoskeleton, spindle, spindle pole. Immunogen sequence: MADQLTEEQI AEFKEAFSLF DKDGDGTITT KELGTVMRSL GQNPTEAELQ DMINEVDADG NGTIDFPEFL TMMARKMKDT DSEEEIREAF RVFDKDGNGY ISAAELRHVM TNLGEKLTDE EVDEMIREAD IDGDGQVNYE EFVQMMTAK
Calmodulin 2 Polyclonal antibody specifically detects Calmodulin 2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
Calmodulin 2 | |
Polyclonal | |
Unconjugated | |
CALM2 | |
1500001E21Rik; AL024017; Calm; Calm1; CALM2; Calm3; CALML2; calmodulin; calmodulin 2; calmodulin 2 (phosphorylase kinase, delta); Calmodulin-1; Calmodulin-2; Calmodulin-3; CAM; CAM1; Cam2; CAM3; CAMB; Camc; CaMI; CaMII; CaMIII; LP7057 protein; LQT15; PHKD; PHKD2; phosphorylase kinase delta; phosphorylase kinase subunit delta; prepro-calmodulin 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
12314, 805 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.08 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P0DP24, P0DP27 | |
CALM2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-149 of human CALM2 (NP_001734.1). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.