missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calsequestrin 1 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-00194AF405
This item is not returnable.
View return policy
Description
Calsequestrin 1 Polyclonal specifically detects Calsequestrin 1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Calsequestrin 1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
844 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
calmitin, calmitine, calsequestrin 1 (fast-twitch, skeletal muscle), Calsequestrin, skeletal muscle isoform, calsequestrin-1, CASQ, PDIB1 | |
Produced in rabbits immunized with purified, recombinant Calsequestrin 1 (P31415; Gln35-Asp396). | |
0.1 mL | |
Primary | |
Human | |
Purified |