Learn More
Tocris Bioscience™ (-)-Cannabidiol
Natural cannabinoid; GPR55 antagonist, weak CB1 antagonist, CB2 inverse agonist and AMT inhibitor
Brand: Tocris Bioscience™ 1570/50
5277.47 DKK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
(-)-Cannabidiol is a non-psychotropic constituent of cannabis that is anticonvulsive, antihyperalgesic and neuroprotective in vivo. GPR55 and weak CB1 antagonist (IC50 values are 0.445 and 3.35 μM), CB2 receptor inverse agonist and inhibitor of anandamide uptake (IC50 = 27.5 μM). Also inhibits FABPs and a weak agonist at TRP vanilloid receptors (EC50 = 3.5 μM).Specifications
2-[(1R,6R)-3-Methyl-6-(1-methylethenyl)-2-cyclohexen-1-yl]-5-pentyl-1,3-benzenediol | |
50 mg | |
C21H30O2 |
13956-29-1 | |
Non-selective Cannabinoid Receptor Antagonists | |
0.99 |