Description
Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
Specifications
Specifications
CAS | 108-48-5, 108-24-7, 109-99-9 |
CAS Max % | 10.00%, 80.00% |
Boiling Point | 66°C.The physical data is that of the main component. |
Quantity | 4 x 2.5 L |
Flash Point | -15°C |
Packaging | Glass Bottle |
Physical Form | Liquid, clear |
Purity Grade Notes | Capping reagents |
Product Content Correction
Product Title
Capping Reagent, Honeywell Burdick & Jackson™ >
Spot an opportunity for improvement?Share a Content Correction