missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals CAPSO, 98% Product Code.: 11449217

Thermo Scientific Chemicals CAPSO, 98%

Product Code. 11449217
100 g, 100g
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11449217

Brand: Thermo Scientific Chemicals B21305.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 73463-39-5 | C9H19NO4S | 237.314 g/mol

CAPSO is used in various biological applications such as electrophoresis. Specific applications include buffers for protein separations and diagnostic reagent production. Also used as biological buffer in the following studies, to maintain pH 9 during water leach test for the analysis of high-carbon coal fly ash-soil mixtures, chemiluminescent in situ hybridization using soybean peroxidase-labeled peptide nucleic acid (PNA) probes, preparation of planar bilayer lipid membranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
CAPSO is used in various biological applications such as electrophoresis. Specific applications include buffers for protein separations and diagnostic reagent production. Also used as biological buffer in the following studies, to maintain pH 9 during water leach test for the analysis of high-carbon coal fly ash-soil mixtures, chemiluminescent in situ hybridization using soybean peroxidase-labeled peptide nucleic acid (PNA) probes, preparation of planar bilayer lipid membranes.

Solubility
Soluble in water (10 g+ 90 ml).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73463-39-5
Molecular Formula C9H19NO4S
Molecular Weight (g/mol) 237.314
MDL Number MFCD00041778
InChI Key INEWUCPYEUEQTN-UHFFFAOYSA-N
Synonym capso, 3-cyclohexylamino-2-hydroxy-1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxypropane-1-sulfonic acid, capso free acid, 1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxy, capso compound, 3-cyclohexylamino-2-hydroxy-1-propanesuhicic acid, 3-cyclohexylamino-2-hydroxy-1-propanesulphonic acid, 3-cyclohexylamino-2-hydroxypropanesulfonic acidShow MoreShow Less
PubChem CID 2733480
IUPAC Name 3-(cyclohexylamino)-2-hydroxypropane-1-sulfonic acid
SMILES C1CCC(CC1)NCC(CS(=O)(=O)O)O

Specifications

Melting Point 270°C to 274°C
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Soluble in water (10 g+90 ml).
Formula Weight 237.32
Percent Purity 98%
Chemical Name or Material CAPSO

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number10250597 Certificate TypeCertificate of Analysis Date30/08/2024 Product CodeB21305.22,B21305.14,B21305.36
Lot Number10248678 Certificate TypeCertificate of Analysis Date09/02/2024 Product CodeB21305.22,B21305.14,B21305.36
Lot Number10246851 Certificate TypeCertificate of Analysis Date07/06/2023 Product CodeB21305.22
Lot Number10159335 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB21305.22,B21305.14,B21305.36
Lot NumberH2310A Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB21305.22,B21305.14,B21305.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals CAPSO, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.