Learn More
Azpack™ Carbon Chrome Steel Spheres
Demonstrates Archimedes' Principle and other properties of physics and math. Azpack™ Carbon Chrome Steel Spheres are used mainly for educational purposes.
Brand: Azpack™ 12423090
This item is not returnable.
View return policy
Description
- Manufactured from carbon chrome steel.
- Available in a variety of sizes
- Smooth, polished chrome surface
- Round-shaped balls
Specifications
Metal Ball | |
Carbon Chrome Steel | |
Dia: 18mm | |
18 mm |
Product Content Correction
Product Title
Azpack™ Carbon Chrome Steel Spheres > 18mm
Spot an opportunity for improvement?Share a Content Correction