Learn More
BUCHI Cartridges
Brand:  BUCHI 044883
Product Code. 12896893
  • 5545.00 DKK / Pack of 50
Estimated Shipment: 11-06-2025
to see stock.
Explore more special offers
Specifications
150 mm
Empty Cartridge
Cartridger C-670 for Packing
Polypropylene
50 frits for packing of 25 cartridges
up to 10 bar
Product Title
BUCHI Cartridges > Dimensions: 40 dia. x 150mmL