Learn More
Memmert™ Castor Frame (2-Part), Height 140 mm, For Un30Pa
Memmert™ Castor Frame for use with Constant Climate Chambers.
Brand: Memmert™ UN30paR9
1551.88 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- 2-parts
- For use with HPP260 Constant Climate Chamber
Specifications
HPP260 | |
Castor Frame | |
140 mm | |
140 mm |
Product Content Correction
Product Title
Memmert™ Castor Frame (2-Part), Height 140 mm, For Un30Pa >
Spot an opportunity for improvement?Share a Content Correction