Learn More
CBFA2T2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11336-1-AP-150UL
 View more versions of this product
Product Code. 16818343
  • 3058.99 DKK / 150µL
This item is not returnable. View return policy
Description
Description
In acute myeloid leukemia, especially in the M2 subtype, the t(8;21)(q22;q22) translocation is one of the most frequent karyotypic abnormalities. The translocation produces a chimeric gene made up of the 5′-region of the RUNX1 (AML1) gene fused to the 3′-region of the CBFA2T1 (MTG8) gene. The chimeric protein is thought to associate with the nuclear corepressor/histone deacetylase complex to block hematopoietic differentiation. The protein encoded by this gene binds to the AML1-MTG8 complex and may be important in promoting leukemogenesis. Several transcript variants are thought to exist for this gene, but the full-length natures of only three have been described.
Specifications
Specifications
CBFA2T2
Polyclonal
Unconjugated
CBFA2T2
CBFA2T2, EHT, MTG8 like protein, MTG8 related protein 1, MTGR1, p85, Protein CBFA2T2, ZMYND3
Rabbit
Antigen Affinity Chromatography
RUO
12396, 296293, 9139
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.8 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O43439, O70374
CBFA2T2
CBFA2T2 Fusion Protein Ag1880
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CBFA2T2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated