Learn More
CD326 (EpCAM) Mouse, anti-Human, Unconjugated, Clone: 323/A3, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60146
Description
- The 323/A3 antibody reacts with the human epithelial cell adhesion molecule (EpCAM or CD326), an ∼40 kDa type I transmembrane glycoprotein which functions as a homophilic and Ca++-independent adhesion molecule
- EpCAM has roles in several cellular processes, including signaling, migration, proliferation, and differentiation
- It is expressed early during erythrogenesis and has been identified as a stem cell marker
- The EpCAM protein is found on the basolateral membrane of most normal epithelial cells and is highly expressed in bone marrow, colon, and lung
- EpCAM is not expressed on mesothelial cells or mesotheliomas, and so is widely used as a diagnostic marker to distinguish mesothelioma and carcinoma cells, as well as serving as a target for immunotherapeutic treatment of carcinomas
Specifications
CD326 (EpCAM) | |
Monoclonal | |
Unconjugated | |
100 μg | |
Human | |
Mammary Cells |
Immunohistochemistry, Flow Cytometry, Immunocytochemistry | |
323/A3 | |
Mouse | |
Primary | |
IgG1 κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.