Learn More
CD37 Antibody (MB-1), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Bio-Techne NBP3-11967C
Product Code. 18310842
  • 4465.33 DKK / 0.10mL
Explore more special offers
This item is not returnable. View return policy
Description
Description
CD37 Monoclonal antibody specifically detects CD37 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
CD37
Monoclonal
DyLight 650
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
MB-1
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
CD37 antigentetraspanin-26, CD37 molecule, cell differentiation antigen 37, GP52-40, leukocyte antigen CD37, MGC120234, Tetraspanin-26, Tspan-26, TSPAN26leukocyte surface antigen CD37
BALB/c mice were immunized with lymph node cells from a patient with nodular poorly differentiated lymphocytic lymphoma. Immunophenotyping studies of these cells identified them as B cells expressing SIgG/lambda, as well as HLA-DR. Leu-12, and B1.
0.1 mL
Cellular Markers, Immunology
951
Store at 4C in the dark.
IgG1 κ