missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RA Mouse anti-Human, APC, Clone: HI100, BD
Mouse Monoclonal Antibody
1960.00 DKK - 3935.00 DKK
Specifications
Antigen | CD45RA |
---|---|
Clone | HI100 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15891429
|
BD Biosciences
561884 |
25 Tests |
1960.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10091934
|
BD Biosciences
550855 |
100 Tests |
3935.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The HI100 monoclonal antibody specifically binds to the 220 kDa isoform of the human leukocyte common antigen, CD45RA. CD45RA is expressed on approximately 40-50% of peripheral CD4+ T cells, 50% of peripheral CD8+ T cells and on a portion of B cells and monocytes. The CD45RA antigen is expressed by naοve and activated T cells. CD45RA-specific antibodies are useful for the study of the suppressor/inducer subpopulation of CD4+ lymphocytes.
Specifications
CD45RA | |
Flow Cytometry | |
APC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
HI100 | |
Monoclonal | |
Mouse | |
Human | |
IgG2b κ | |
Affinity Purified |
For Research Use Only.