missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CD58 (Human) Recombinant Protein
Human CD58 full-length ORF ( AAH05930, 1 a.a. - 240 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000965-P01.25ug
This item is not returnable.
View return policy
Description
- Sequence: MVAGSDAGRALGVLSVVCLLHCFGFISCFSQQIYGVVYGNVTFHVPSNVPLKEVLWKKQKDKVAELENSEFRAFSSFKNRVYLDTVSGSLTIYNLTSSDEDEYEMESPNITDTMKFFLYVLESLPSPTLTCALTNGSIEVQCMIPEYYNSHRGLIMYSWDCPMEQCKRNSTSIYFKMENDLPQKIQCTLSNPLFNTTSSIILTTCIPSSGHSRHRYALIPIPLAVITTCIVLYMNGMYAF
Specifications
AAH05930 | |
CD58 molecule | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD58 | |
GST |
965 | |
Wheat germ expression system | |
25 μg | |
LFA-3, LFA3 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |