Learn More
CD59 Monoclonal Antibody (M011), Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen MA547653
Product Code. 17876818
  • 3712.00 DKK / 100µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
Clone M011 detects 15-20 kDa* bands corresponding to CD59 on SDS-PAGE immunoblots of native human MDA-MB-231, A549, A431, and MCF7 cell lysates, as well as recombinant human CD59 protein. The antibody does not detect denatured CD59. The antibody can be used for multiple applications including ELISA capture, western blot, immunocytochemical labeling and immunoprecipitation. In addition, the antibody labels live, unfixed A549 and MeWo cells.
CD59 (Protectin) is a small (18-20 kDa) GPI-anchored ubiquitously expressed inhibitor of the membrane attack complex (MAC). CD59 is the key regulator that preserves the autologous cells from terminal effector mechanism of the complement cascade. CD59 also associates with C5b-8 complex and counteracts appropriate formation of cytolytic pore within the plasma membrane. Further, CD59 is a low-affinity ligand of human CD2, causes T cell costimulation, and is involved in lymphocyte signal transduction. CD59 is a potent inhibitor of the complement membrane attack complex, whereby it binds complement C8 and/or C9 during the assembly of this complex, thereby inhibiting the incorporation of multiple copies of C9 into the complex, which is necessary for osmolytic pore formation. Mutations in this gene cause CD59 deficiency, a disease involving hemolytic anemia, thrombosis, and cerebral infarction. Multiple alternatively spliced transcript variants of CD59 have been identified.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD59
Monoclonal
Lot-Specific
PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
P13987
CD59
Clone M011 was generated from a proprietary antigen related to the mature form of human CD59 from the MDA-MB-231 breast carcinoma cell line.
100 μL
Primary
Human
Liquid
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
M011
Unconjugated
CD59
16.3A5; 1F5; 1F5 antigen; 20 kDa homologous restriction factor; AA987121; Cb59b molecule; CD59; CD59 antigen; CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5, EJ16, EJ30, EL32 and G344); CD59 glycoprotein; CD59 molecule; CD59 molecule (CD59 blood group); CD59 molecule, complement regulatory protein; Cd59a; CD59a antigen; CD59A glycoprotein; Cd59b; CD59b antigen; CD59B glycoprotein; CD59b moleucle, complement regulatory protein; complement inhibitor CD59; complement regulatory protein; EJ16; EJ30; EL32; G344; HRF20; HRF-20; human leukocyte antigen MIC11; Ly-6-like protein; lymphocytic antigen CD59/MEM43; MACIF; MAC-inhibitory protein; MACIP; MAC-IP; MEM43; MEM43 antigen; membrane attack complex (MAC) inhibition factor; membrane attack complex inhibition factor; Membrane inhibitor of reactive lysis; MIC11; MIN1; MIN2; MIN3; MIRL; MSK21; p18-20; protectin; surface anitgen recognized by monoclonal antibody 16.3A5; T cell-activating protein
Mouse
Protein G
RUO
966
-20° C, Avoid Freeze/Thaw Cycles
IgG1