Learn More
Abnova™ CDK4 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Brand:  Abnova™ H00001019-T02.100uL
Product Code. 16146104
  • 1710.00 DKK / 100µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
293T
CDK4 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 33.7kDa
Transient overexpression cell lysate was tested with anti-CDK4 antibody (Mfr. No. H00001019-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
NM_000075.2
pCMV-CDK4 full-length
100 μL
Human
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ CDK4 293T Cell Transient Overexpression Lysate (Denatured) > 100μL