Learn More
CDK7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534791
Description
Recommended positive controls: 293T, A431, A375, Neuro2A, GL261, C8D30, NIH-3T3, BCL-1, Raw264.7, C2C12. Predicted reactivity: Mouse (95%), Zebrafish (85%), Xenopus laevis (86%), Pig (97%), Rhesus Monkey (99%), Bovine (96%). IHC notes, Requires antigen retrieval using heat mediated 10mM Citrate buffer (pH6.0) or Tris-EDTA buffer (pH8.0) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The trimeric complex of CDK7, cyclin H and MNAT1, functions as a Cdk-activating kinase (CAK). It is an essential component of the transcription factor TFIIH, that is involved in transcription initiation and DNA repair. This complex is thought to serve as a direct link between the regulation of transcription and the cell cycle. The bound complex is important in the normal double-strand break repair function of breast cancer, type 1 (BRCA1). CDK7 may be a target of germline cancer-inducing mutations.
Specifications
CDK7 | |
Polyclonal | |
Unconjugated | |
CDK7 | |
39 KDa protein kinase; 39 protein kinase; AI323415; AI528512; C230069N13; CAK; CAK1; CDK; CDK7; CDK-activating kinase; CDK-activating kinase 1; Cdkn7; cell division protein kinase 7; CR4 protein kinase; CRK4; CRK4 PK (CDC2-related-kinase-4 protein kinase); cyclin dependent kinase 7; cyclin-dependent kinase 7; cyclin-dependent kinase 7 (homolog of Xenopus MO15 cdk-activating kinase); cyclin-dependent kinase 7 (MO15 homolog Xenopus laevis cdk-activating kinase); cyclin-dependent kinase 7 (MO15 homolog, Xenopus laevis, cdk-activating kinase); EC 2.7.11.22; EC 2.7.11.23; HCAK; homolog of Xenopus MO15 Cdk-activating kinase; kinase subunit of CAK; MO15; Mpk-7; OTTHUMP00000222032; OTTHUMP00000222033; p39 Mo15; p39MO15; Protein-tyrosine kinase MPK-7; serine/threonine kinase stk1; serine/threonine protein kinase 1; serine/threonine protein kinase MO15; Serine/threonine-protein kinase 1; STK1; TFIIH basal transcription factor complex kinase subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1022, 12572 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal, 0.025% ProClin 300; pH 7 | |
P50613, Q03147 | |
CDK7 | |
Full length human CDK7 Recombinant protein. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.