missing translation for 'onlineSavingsMsg'
Learn More

CDw75 Ab-1 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments with Epredia™ CDw75 Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-130-P

 View more versions of this product

Product Code. 12619536

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

CDw75 is a neuraminidase-sensitive sialoprotein, present on cell membrane and cytoplasm of germinal center B-cells and derived lymphomas. LN1 reacts with RBC precursors of bone marrow, ductal and ciliated epithelial cells of kidney, breast, prostate, pancreas, lung, and with glioblastomas, astrocytomas, and Reed Sternberg cells in lymphocyte predominant Hodgkin's disease. LN1 is shown to be a reliable antibody for ascribing a B-cell phenotype in known lymphoid tissues.

Host Species: Mouse

Clone: LN1

Isotype: IgM/κ

Species Reactivity: Human. Does not react with rat. Others not known

Epitope: Not determined

Immunogen: Nuclei from pokeweed mitogen-stimulated peripheral blood lymphocytes

Molecular Weight: 53 and 87 kDa

Positive Control: Lymph node or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry
Specifications

Specifications

CDw75 Ab-1
Monoclonal
200μg/mL
Mouse
1 mL
B-Cell
Human
IgM κ
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
LN1
Unconjugated
Nuclei from pokeweed mitogen-stimulated peripheral blood lymphocytes
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.