Learn More
Lonza™ Cell Line Optimization 4D-Nucleofector™ X Kit
Brand:  Lonza™ V4XP-9096
 View more versions of this product
Product Code. 13409339
  • 16570.00 DKK / 96 reactions
Explore more special offers
This item is not returnable. View return policy
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4°C
4D-Nucleofector™ X Unit
96
1 Each
pmaxGFP™
Includes 20μL Transfection volume, 0.675mL Nucleofector Solution, 4 x 0.15mL Supplement, 2x 50μg pmaxGFP Vector, 2 Single Nucleocuvette (100 μl), Store at 4°C, For Use with 4D-Nucleofector X Unit
Cell Line Optimization 4D-Nucleofector X Kit