Learn More
Thermo Scientific™ Cell Locker™ Chamber with Copper Tray, Stainless Steel
Protected chambers for your most sensitive cells
Brand: Thermo Scientific™ 50154739
7482.91 DKK valid until 2025-03-31
Use promo code "24119" to get your promotional price.
This item is not returnable.
View return policy
Description
Protected chambers for your most sensitive cells
- Each individual Cell Locker Chamber is effectively a quarantine chamber.
- Dual 0.2μm membrane filters permits air circulation but exclude microbial contaminants.
- The replacable 0.2μm membrane filter is hydrophobic, oleophobic, resistant to organic solvents and tested for biosafety and low cytotoxicity.
- Independent 3rd party tests validate protection against circulating microorganisms (when used as directed)
Each Cell Locker Chamber holds 9 each T-75 cell culture flasks, 20 each 6-well plates, or 24 each 96-well plates.
- Dishes and flasks can be removed individually on a work tray or inside the entire Cell Locker Chamber with optional transport cover, thus protecting samples from outside air.
- The non skid tray slides out to allow access to culture vessels, or the entire tray can be removed.
- The Cell Locker Chamber can be manually disinfected or can be autoclaved.
Warranty: 1 year
Specifications
Cell Locker Chamber with copper tray | |
9 each T-75 cell culture flasks, 20 each 6-well plates, or 24 each 96- well plates | |
Stainless Steel | |
1 | |
1 Year |
Stainless Steel | |
LED | |
53 | |
Accessory |
Brochure: Cell Locker VIOS 160i CO2 Incubator NA
File size: 1.3 MB
Brochure: Cell Locker VIOS 160i CO2 Incubator EU
File size: 1.3 MB
Brochure: Cell Locker Forma Steri-cycle CO2 Incubator
File size: 1.3 MB
Special Offers
- Multiple Manufacturers
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Product Content Correction
Product Title
Thermo Scientific™ Cell Locker™ Chamber with Copper Tray, Stainless Steel >
Spot an opportunity for improvement?Share a Content Correction