Learn More
Greiner Bio-One CELLSTAR Suspension Cell Culture Flasks
GREENER_CHOICE
Designed to culture cell suspended in a growth medium. Greiner Bio-One™ CELLSTAR Suspension Cell Culture Flasks are manufactured from polystyrene for optimum visibility and strength.
Brand:  Greiner Bio-One 658190
 View more versions of this product
Product Code. 10116520
  • 1501.91 DKK / Pack of 120
Only null left
Estimated Shipment: 04-08-2025
Description
  • Hydrophobic surface, ideal for suspension cultures and hybridoma
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic, non-cytotoxic
  • Graduation on the side
  • Canted neck
  • Easily distinguished by their white screw caps
  • White standard screw cap with ventilation position
  • TC surface treatment: -/suspension
  • Sterile and user friendly packaging
TRUSTED_SUSTAINABILITY
Specifications
Specifications
White
Polystyrene
120
Screw
250 mL
120 Pack
Yes
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Greiner Bio-One CELLSTAR Suspension Cell Culture Flasks > Screw Cap Type: White Standard Screw Cap; Capacity: 250mL