Learn More
Eppendorf™ CellXpert™ C170i, Segmented Inner Door
CellXpert™ C170i CO2 incubator provides reliable growth conditions for more reproducible results.
Brand: Eppendorf™ 6731000041
86288.62 DKK valid until 2025-06-30
Use promo code "24075" to get your promotional price.
View more versions of this product
Description
- Advanced user Interface and front USB-ports
- Fan-less design, smart racking system and innovative insulation concept
- New magnetic door locking concept for outer and inner doors
- Seamless chamber, easy disassembling
- 180°C High Temperature Disinfection (HTD) with protocol, no internal HEPA-filters
- Customizable tasks and alarms
- Standard building management system and VisioNize connected lab
- Vibration and turbulence protection
Specifications
170 L | |
IR Sensor | |
Ambient +4° to 50°C | |
Right-Sided Handle | |
2 | |
71.5 x 71.8 x 90.0 cm |
Stainless Steel | |
EU | |
VisioNize™ touch interface | |
ZrO2 (zirconium dioxide) | |
44.5 x 53.9 x 69.2 cm | |
220 to 240 V, 50/60 Hz |
Product Suggestions
Customers who viewed this item also viewed.
Videos
-
Eppendorf CellXpert C170i CO2 Incubators
-
Eppendorf CellXpert C170i CO2 Incubators - For More Reproducible Cell Culture Results
-
Eppendorf CellXpert C170i CO2 Incubators
-
Eppendorf CellXpert C170i CO2 Incubators - For More Reproducible Cell Culture Results
Promotions
-
Multiple Manufacturers
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.Promo Code: 24075Expires: 2025-06-30
Days
12Hours
8Minutes
43Seconds
49New Offer Save Up to 78%