Learn More
Eppendorf CellXpert™ CS220
GREENER_CHOICE
CO2 incubator shaker with an integrated 180°C sterilization and an outstanding capacity-to-footprint ratio.
337500.00 DKK
Specifications
Model CellXpert CS220
View More Specs
Products 2
Product Code Brand Speed Range Price Quantity & Availability  
Product Code Brand Speed Range Price Quantity & Availability  
30213522
GREENER_CHOICE
View Documents
Eppendorf
6740211060
20 to 300 rpm
337500.00 DKK
Each
30213519
GREENER_CHOICE
View Documents
Eppendorf
6740212060
20 to 250 rpm
337500.00 DKK
Each
Description
Description
  • CO2 incubator shaker
  • Stackable up to 3-fold
  • Non-refrigerated
  • 8°C above ambient to 60°C
  • Accessories available to purchase separately: platform, clamps, sticky pads, storage base or feet, water supply package, 1-2 stacking kits and water tank holder
Specifications
Specifications
CellXpert CS220