Learn More
Eppendorf CellXpert™ CS220
CO2 incubator shaker with an integrated 180°C sterilization and an outstanding capacity-to-footprint ratio.
337500.00 DKK
Product Code | Brand | Speed Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Speed Range | Price | Quantity & Availability | |||||
30213522
|
Eppendorf
6740211060 |
20 to 300 rpm |
337500.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30213519
|
Eppendorf
6740212060 |
20 to 250 rpm |
337500.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- CO2 incubator shaker
- Stackable up to 3-fold
- Non-refrigerated
- 8°C above ambient to 60°C
- Accessories available to purchase separately: platform, clamps, sticky pads, storage base or feet, water supply package, 1-2 stacking kits and water tank holder
Specifications
CellXpert CS220 |